Få mere at vide
Sildenafil Chlorosulfonyl (5-(5-Chlorosulfonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one), Mikromol™
Artikelnummer. 30049486
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049486 25 mg
30052280 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049486 Leverandør Mikromol Leverandørnr. MM0254.340025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sildenafil Chlorosulfonyl (5-(5-Chlorosulfonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(5-Chlorosulfonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
CAS 139756-22-2
Analyt- eller komponentnavne 5-(5-Chlorosulfonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C17 H19 Cl N4 O4 S
Certificeringer/compliance ISO 17025
Synonym Benzenesulfonyl chloride, 3-(6,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxy-, Benzenesulfonyl chloride, 3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxy- (9CI), 1H-Pyrazolo[4,3-d]pyrimidine, benzenesulfonyl chloride deriv., 5-(5-Chlorosulfonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
InChI formel InChI=1S/C17H19ClN4O4S/c1-4-6-12-14-15(22(3)21-12)17(23)20-16(19-14)11-9-10(27(18,24)25)7-8-13(11)26-5-2/h7-9H,4-6H2,1-3H3,(H,19,20,23)
SMIL CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)Cl
IUPAC navn 4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonyl chloride
Molekylvægt (g/mol) 410.88
Mængde 25 mg
Formel vægt 410.0816 g/mol