Få mere at vide
BRAND™ SILBERBRAND™ Borosilicate Glass Automatic Burette with Bottle
Shop alle Brand produkter
Kapacitet (metrisk):
25 ml
Pakningsstørrelse:
Hver
Beskrivelse
- Borosilicate glass (Boro 3.3) construction
- Automatic burette
- Pellet pattern
- Includes lateral needle-valve stopcock with PTFE-key
- Includes bottle
- Class B
- Schellbach stripe
- Graduations in black
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 25 ml |
| Materiale | Borosilicate Glass |
| Gradueringer | 0.05 mL |
| Inkluderer | Schellbach Stripe |
| Stophane materiale | PTFE Key |
| Tolerance | ±0.05 mL |
| Type | Automatic Buret |
| Klasse | Class B |
| Certificeringer/compliance | DIN EN ISO 385 |