Learn More
Endecotts Sieve Lid 203
Product Code. 11532015
Change view
Click to view available options
For Use With (Equipment):
For use with Sieves
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11532015 For use with Sieves Each
1 options
Product Code. 11532015 Supplier Endecotts Supplier No. 008S/STLID
  • ONLINE EXCLUSIVE PRICE
    455.00 DKK
    313.20 DKK / Pakke med 1
    Spare 141.80 DKK
    31% Slukket
 Restricted Item
Forventet forsendelse: 14-01-2027
Stainless Steel Sieve Lid
  • Stainless steel sieve lid designed for Sieve 203, providing secure coverage during sieving operations and supporting clean, controlled, and reliable sample handling in laboratory and testing environments.
Specifications
Product Type Lid
Diameter (English) 8 in.
Diameter (Metric) 203 mm
For Use With (Equipment) For use with Sieves