Få mere at vide
R&D Systems™ SIB 1508Y maleate
Beskrivelse
SIB 1508Y maleate is a potent neuronal nicotinic ACh receptor agonist (EC50 values are 1.8, 5, 9 and 23 nM for α4β2, α2β4, α4β4 and α3β4 receptors respectively). Improves cognitive function in a MPTP-induced model of Parkinson's Disease in vivo.
Tekniske data
Tekniske data
| Renhed | >98% |
| Mængde | 50 mg |
| Produkttype | SIB 1508Y maleate |
| Molekylvægt (g/mol) | 302.33 |
| Produktlinje | Tocris |
| Inhibitorer | SIB 1508Y maleate |
For Research Use Only