Learn More
Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP)
Shrimp Alkaline Phosphatase (SAP) is a high specific activity, heat-labile alkaline phosphatase purified from a recombinant source and originally isolated from Pandalus borealis (arctic shrimp).
Brand: Applied Biosystems™ 783905000UN
Additional Details : Weight : 21.59000kg
Description
- 100% heat-inactivated in 5 min at 65°C
- Significantly improved storage stability at lower temperatures
- Very high specific activity
- Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
- Purified from a recombinant source
- May be added directly to restriction enzyme digests
- No vector purification necessary
- Requires no supplemental zinc or other additives for activity
- Works direct in many different buffers
- Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Order Info
5000 Units
Specifications
Alkaline Phosphatase | |
Dephosphorylation of DNA/RNA | |
Shrimp Alkaline Phosphatase, 5000 Units, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases | |
5000 Units |
For Research Use Only. Not for use in diagnostic procedures.