missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP)
Shrimp Alkaline Phosphatase (SAP) is a high specific activity, heat-labile alkaline phosphatase purified from a recombinant source and originally isolated from Pandalus borealis (arctic shrimp).
459.00 DKK - 3285.00 DKK
Specifications
Enzyme | Alkaline Phosphatase |
---|---|
For Use With (Application) | Dephosphorylation of DNA/RNA |
Product Code | Brand | Item Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Item Description | Quantity | Price | Quantity & Availability | ||||
15859906
|
Applied Biosystems™
783901000UN |
Shrimp Alkaline Phosphatase, 1000 Units, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases | 1000 Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15879906
|
Applied Biosystems™
783905000UN |
Shrimp Alkaline Phosphatase, 5000 Units, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases | 5000 Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15889906
|
Applied Biosystems™
78390500UN |
Shrimp Alkaline Phosphatase, 500 Units, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases | 500 Units |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- 100% heat-inactivated in 5 min at 65°C
- Significantly improved storage stability at lower temperatures
- Very high specific activity
- Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
- Purified from a recombinant source
- May be added directly to restriction enzyme digests
- No vector purification necessary
- Requires no supplemental zinc or other additives for activity
- Works direct in many different buffers
- Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Order Info
500 Units
Specifications
Alkaline Phosphatase | |
Dephosphorylation of DNA/RNA |
For Research Use Only. Not for use in diagnostic procedures.