Få mere at vide
Shikimic Acid Ethyl Ester, TRC
Artikelnummer. 30132680
Skift visning
Mængde:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132680 2.5 g
30137874 1 g
30004522 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132680 Leverandør Toronto Research Chemicals Leverandørnr. TRCS3571052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 101769-63-5
Molekylær formel C9H14O5
Molekylvægt (g/mol) 202.2
Synonym (3R,4S,5R)-Trihydroxy-1-cyclohexene-1-carboxylic Acid Ethyl Ester;[3R-(3alpha,4alpha,5beta)]-Trihydroxy1-Cyclohexene-1-carboxylic Acid Ethyl Ester, Ethyl Shikimate
IUPAC navn ethyl (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMIL CCOC(=O)C1=C[C@@H](O)[C@@H](O)[C@H](O)C1
Tekniske data
Kemisk navn eller materiale Shikimic Acid Ethyl Ester
Mængde 2.5 g
InChI formel InChI=1S/C9H14O5/c1-2-14-9(13)5-3-6(10)8(12)7(11)4-5/h3,6-8,10-12H,2,4H2,1H3/t6-,7-,8-/m1/s1
Formel vægt 202.0841