missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, FITC, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-75352
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunomicroscopy
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| FITC | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free | |
| This Sheep anti-Rabbit IgG (H+L) Secondary Antibody [FITC] was developed against purified rabbit IgG (H&L). | |
| 2 mg | |
| Adaptive Immunity, Immunology | |
| Rabbit | |
| Purified |
| Flow Cytometry, Immunomicroscopy | |
| 1.5 mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Sheep | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion