Få mere at vide
Sheep anti-Mouse Kappa Light Chain Secondary Antibody, TRITC, Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Tekniske data
Tekniske data
| Antigen | Kappa Light Chain |
| Applikationer | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Klassifikation | Polyclonal |
| Konjugeret | TRITC |
| Fortynding | ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Formulering | Lyophilized from PBS (pH 7.2) |
| Gene Alias | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Værtsarter | Sheep |
| Immunogen | A pool of purified Bence Jones kappa proteins isolated from mouse urine. Freund's complete adjuvant is used in the first step of the immunization procedure. |
| Oprensningsmetode | Ion exchange chromatography |
| Vis mere |