missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Sheep anti-Bovine IgG2 Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB786-0.5mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG2 Heavy Chain Polyclonal antibody specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| IgG2 Heavy Chain | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| 50 mM HEPES pH 7.1, 0.1 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2 | |
| Bovine IgG2 | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:1000 - 1:10000, ELISA 1:1000- 1:10000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Bovine | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion