missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Sheep anti-Bovine IgG Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB791-0.5mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| IgG Heavy Chain | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| 50 mM HEPES pH 7.1, 0.1 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2 | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:5000- 1:50000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
| Immunoglobulin G, ImmunoglobulinG | |
| Bovine IgG-heavy chain | |
| 0.5 mg | |
| Immunology | |
| Bovine | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion