Beskrivelse
- Acetal polymer construction
- Small slot-style pores
- Cassette writing face is at a 45° angle
- Box of 500 cassettes with attached lids
Tekniske data
Tekniske data
| Pore stil | Small Slotted Pore |
| Skriveansigtsvinkel | 45° |
| Farve | Lyserød |
| Materiale | Acetal Polymer |
| Produkttype | Tissue Cassette |