Få mere at vide
Epredia™ Shandon™ Scalpel Blade No. 21, Sterile, 2 in. (5.1cm)
Artikelnummer. 12657776 Shop alle Epredia produkter
Størrelse:
# 21
Pakningsstørrelse:
Pakke med 100
Artikelnummer. 12657776
Brand: Epredia™ 53221
Cut precisely with the Epredia™ Shandon™ Scalpel Blade No. 21, which is 2.06 in. (5.1cm) in length.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Tekniske data
Størrelse # 21
Længde (engelsk) 2 in.
Længde (metrisk) 5.1 cm
Sterilitet Sterile
Mængde 10/Pk.