Learn More
Epredia™ Shandon™ Premium High-profile Disposable Blades
Choose either PTFE-coated or non-coated Epredia™ Shandon™ premium heavy-duty high-profile disposable blades.
Brand: Epredia™ 1001593
884.37 DKK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 0.45360kg
Specifications
Premium Microtomy Blade | |
High | |
Yes | |
PTFE |