Beskrivelse
Achieve thorough tissue infiltration and provide maximum support during microtomy with high-quality Thermo Scientific™ Shandon™ Paraffin.
- Quality blend of paraffin, polymers and plastics (to varying degrees, based upon type)
Tekniske data
Tekniske data
| Smeltepunkt | 50° to 54°C |
| Produkttype | Paraffin |
| Mængde | 8 x 1kg |
| Til brug med (applikation) | Thin sectioning |