Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Product Code. 10446640 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cell Preparation System
Replacement Reagent
Unit Size:
1 pair
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10446640 Replacement Reagent Each
10376294 Cell Preparation System 1 pair
10066588 Cell Preparation System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10446640 Supplier Epredia™ Supplier No. 7401151
  • ONLINE EXCLUSIVE PRICE
    465.00 DKK
    350.53 DKK / Pakke med 1
    Spare 114.47 DKK
    24% Slukket
View all promotions
 Restricted Item
Forventet forsendelse: 18-12-2026
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Reagent