missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ SH3PXD2A Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142568
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.
Tekniske data
| SH3PXD2A | |
| Polyclonal | |
| Unconjugated | |
| Sh3pxd2a | |
| 2310014D11Rik; AA589508; Adapter protein TKS5; adaptor protein TKS5; AI256723; AI413738; C230050L11; EG329070; FISH; Five SH3 domain-containing protein; five SH3 domains; Gm5098; KIAA0418; SH3 and PX domain-containing protein 2A; SH3 and PX domains 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Sh3md1; SH3PXD2A; Tks5; TSK5; Tyrosine kinase substrate with five SH3 domains | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9644 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5TCZ1 | |
| Sh3pxd2a | |
| Peptide with sequence C-KPIEKSQFIHNNLK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion