Learn More
Invitrogen™ Human SGKL (SGK3), GST Tag Recombinant Protein
Human SGKL (SGK3), GST Tag Recombinant Protein
Brand: Invitrogen™ PR7301A
Additional Details : Weight : 0.01000kg
Description
Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK.
Specifications
NP_037389, S487D | |
74.3 kDa | |
SGKL (SGK3) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: AGC Group | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
100μg | |
SGK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |