Learn More
Trajan™ GC Stainless Steel Nuts
Provide optimum system connections for SGE fittings.
Brand:  Trajan™ 103403
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15209113
  • 510.00 DKK / Pack of 5
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • For use with SGE fittings.
  • Stainless Steel Construction
Specifications
Specifications
Nut
1/16 in.
Stainless Steel
SGE Normal or Short Fittings