Learn More
Trajan™ Diamond MS CTC PAL Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™ 004770
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13177976
  • 860.00 DKK / Each
Estimated Shipment: 02-07-2024
to see stock.
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Gas/Liquid chromatography
662°F
50R-CTC-GT-MS1
50 μL
Removable
Diamond MS Syringe
350°C
Ea.
CTC PAL Autosampler Syringe