missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes

Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.

Brand:  Trajan™ 0355220

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13157906

  • 653.00 DKK / Each

Estimated Shipment
11-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Improve the sensitivity of your MS analysis:

  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications

Specifications

With Ultra Smooth Hydrophobic coating
0.006 in.
0.17 mm
Ultra smooth hydrophobic coating
Needle
LC
22 G
0.028 in.
0.72 mm
2.007 in.
51mm
10 to 50μL manual and autosampler syringe
Videos
SDS
Documents