Learn More
Trajan™ Diamond GC Needles for 100 to 500μL Manual Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 100 to 500μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
Brand:  Trajan™ 0355332
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13147926
  • 675.00 DKK / Each
Estimated Shipment: 11-07-2024
to see stock.
Description
Description
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
With Ultra Smooth coating
0.007 in.
0.2 mm
2 in.
Bevel
25 G
0.0196 in.
0.49 mm
GC Needle
Manual Syringe (100 to 500μL)