Learn More
Trajan™ 0.5μL Plunger-in-Needle Syringes
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
Brand: Trajan™ 033620
Additional Details : Weight : 0.50000kg
Description
- Plunger extends into the needle tip
- Sample is contained only in the needle, i.e. no glass contact
- Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
Borosilicate Glass (Barrel) | |
0.003 in. | |
0.009 in. | |
8 mm | |
For nanovolume capillary chromatography injection | |
Replacement Plunger | |
75 mm |
23 G | |
0.10 mm | |
0.31 in. | |
0.23 mm | |
2.95 in. | |
0.5 μL | |
Cone |