missing translation for 'onlineSavingsMsg'
Få mere at vide

SGC0946, MedChemExpress

Artikelnummer. 16499267
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16499267 5 mg
16486718 10 mg
16402807 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16499267 Leverandør Medchem Express Leverandørnr. HY15650

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress SGC0946 is a selective DOT1LH3K79 methyltransferase) inhibitor, with an IC50 of 0.3 nM. SGC0946 results in G1 arrest, inhibits potential of cell self-renewal and metastatic, also induces cell differentiation. SGC0946 can be used in studies of leukemia and solid tumors and also serve as a probe to further investigate the cellular mechanism of DOT1L in both normal and diseased cells.

SGC0946 is a selective DOT1LH3K79 methyltransferase) inhibitor, with an IC50 of 0.3 nM. SGC0946 results in G1 arrest, inhibits potential of cell self-renewal and metastatic, also induces cell differentiation. SGC0946 can be used in studies of leukemia and solid tumors and also serve as a probe to further investigate the cellular mechanism of DOT1L in both normal and diseased cells.

Tekniske data

Kemisk navn eller materiale SGC0946
CAS 1561178-17-3
Farve Light Nude
Renhedsgrad noter Research
Molekylær formel C28H40BrN7O4
Mængde 5 mg
Opløselighedsinformation DMSO : 50 mg/mL (80.83 mM; Need ultrasonic)
SMIL NC1=NC=NC2=C1C(Br)=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](CN(CCCNC(NC4=CC=C(C(C)(C)C)C=C4)=O)C(C)C)O3
Molekylvægt (g/mol) 618.57
Formel vægt 618.57
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.