Få mere at vide
Novus Biologicals™ SFXN2 Overexpression Lysate
Artikelnummer. 18158309 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18158309 0.1 mg
1 options
Artikelnummer. 18158309 Leverandør Novus Biologicals™ Leverandørnr. NBL115899
 Begrænset vare
Forventet forsendelse: 09-10-2026
The SFXN2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC2148 and based on accession number NM_178858. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens sideroflexin 2 (SFXN2), mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only