missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SF-1/NR5A1/Steroidogenic Factor 1 Antibody (SF1/8150R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20855JF646
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| SF-1/NR5A1/Steroidogenic Factor 1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
| Recombinant fragment (around aa200-400) of human SF-1/NR5A1/Steroidogenic Factor 1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology | |
| 2516 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SF1/8150R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion