Få mere at vide
Sevelamer Hydrochloride (Technical Grade), TRC
Artikelnummer. 30138539
Skift visning
Mængde:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138539 5 g
30138019 500 mg
17972355 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138539 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2560005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 152751-57-0
Molekylær formel (C3H7N)m• (C3H5ClO)n• (HCl)x
Molekylvægt (g/mol) 186.08
Synonym 2-Propen-1-amine Hydrochloride polymer with 2-(Chloromethyl)oxirane, 2-Propen-1-amine Hydrochloride polymer with (Chloromethyl)oxirane, (Chloromethyl)oxirane polymer with 2-Propen-1-amine Hydrochloride, Allylamine Hydrochloride-epichlorhydrin Copolymer, Allylamine Hydrochloride-epichlorohydrin Copolymer, GT 16-026A, Phosblock, RenaGel;
SMIL NCC=C.ClCC1OC1
Tekniske data
Kemisk navn eller materiale Sevelamer Hydrochloride
Mængde 5 g
InChI formel InChI=1S/C3H5ClO.C3H7N.ClH/c4-1-3-2-5-3;1-2-3-4;/h3H,1-2H2;2H,1,3-4H2;1H