Få mere at vide
Setiptiline-d3, TRC
Artikelnummer. 30422609
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30422609 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422609 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2808021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Setiptiline-d3
CAS 1795024-97-3
Molekylær formel C19H16D3N
Mængde 1 mg
Synonym 2,3,4,9-Tetrahydro-2-(methyl-d3)-1H-dibenzo[3,4:6,7]cyclohepta[1,2-c]pyridine, Org 8282-d3, MO-8282-d3, Teciptilline-d3
InChI formel InChI=1S/C19H19N/c1-20-11-10-18-16-8-4-2-6-14(16)12-15-7-3-5-9-17(15)19(18)13-20/h2-9H,10-13H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N(C1)CCC2=C1C(C=CC=C3)=C3CC4=C2C=CC=C4
Molekylvægt (g/mol) 264.38
Formel vægt 264.1706