Få mere at vide
Setanaxib, MedChemExpress
Artikelnummer. 16457797
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16457797 5 mg
16455078 10 mg
16451036 200 mg
16421317 50 mg
16405816 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16457797 Leverandør Medchem Express Leverandørnr. HY12298
MedChemExpress Setanaxib (GKT137831) is a selective NADPH oxidase (NOX1/4) inhibitor with Kis of 140 and 110 nM, respectively.
Setanaxib (GKT137831) is a selective NADPH oxidase (NOX1/4) inhibitor with Kis of 140 and 110 nM, respectively.
Tekniske data
Kemisk navn eller materiale Setanaxib
Fysisk form Powder
CAS 1218942-37-0
Farve Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C21H19ClN4O2
Til brug med (applikation) COVID-19-immunoregulation
Synonym GKT137831 GKT831
Opløselighedsinformation DMSO : 14.29 mg/mL (36.19 mM; Need ultrasonic)
SMIL O=C1N(C2=CC=CC=C2Cl)NC(C1=C(C3=CC=CC(N(C)C)=C3)N4C)=CC4=O
Molekylvægt (g/mol) 394.85
Formel vægt 394.85
Procent renhed 95.0%