Learn More
Gilson™ Maintenance Kit for Pipetman™
Manufactured by Gilson™. Gilson™ Maintenance Kit for Pipetman™ are guaranteed to deliver value and assure reliability and precision.
Brand:  Gilson™ F144502
Additional Details : Weight : 0.00720kg
Product Code. 10377132
  • 621.00 DKK / Each
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • Maintenance kit  
Specifications
Specifications
For Pipetman: P10
1 Tip-holder, 3 piston seals and 3 O-rings