Learn More
SERVALYT™ 7-9 Carrier Ampholytes, Serva Electrophoresis™
Brand:  Serva Electrophoresis 4290704
Encompass_Preferred
Quantity 2 mL
Product Code. 11782437
  • 514.54 DKK / Each
Only null left
Estimated Shipment: 07-07-2025
Specifications
Liquid
Brown
Uncharacteristic
SERVALYT 7-9
2 mL
Fully miscible
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SERVALYT™ 7-9 Carrier Ampholytes, Serva Electrophoresis™