Få mere at vide
Sertindole-d4, TRC
Artikelnummer. 30410920
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30410920 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410920 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2795021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sertindole-d4
CAS 1794737-42-0
Molekylær formel C24H22D4ClFN4O
Mængde 1 mg
Synonym 1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl]-1-piperidinyl]ethyl-d4]-2-imidazolidinone, Serdolect-d4, Zerdol-d4
InChI formel InChI=1S/C24H26ClFN4O/c25-18-1-6-23-21(15-18)22(16-30(23)20-4-2-19(26)3-5-20)17-7-10-28(11-8-17)13-14-29-12-9-27-24(29)31/h1-6,15-17H,7-14H2,(H,27,31)/i13D2,14D2
SMIL ClC1=CC2=C(N(C3=CC=C(F)C=C3)C=C2C4CCN(C([2H])([2H])C([2H])([2H])N5C(NCC5)=O)CC4)C=C1
IUPAC navn 1-[2-[4-[5-chloro-1-(4-fluorophenyl)indol-3-yl]piperidin-1-yl]-1,1,2,2-tetradeuterioethyl]imidazolidin-2-one
Molekylvægt (g/mol) 444.97
Formel vægt 444.203