Få mere at vide
Sertaconazole Nitrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sertaconazole Nitrate |
| CAS | 99592-39-9 |
| Analyt- eller komponentnavne | Sertaconazole Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H15 Cl3 N2 O S . H N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Sertaconazole nitrate, (RS)-1-[2-[(7-Chloro-1-benzothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate, 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, mononitrate (9CI), Dermofix, Dermovit, Ertaczo, FI 7056, Sertaconazole mononitrate, Zalain |
| InChI formel | InChI=1S/C20H15Cl3N2OS.HNO3/c21-14-4-5-16(18(23)8-14)19(9-25-7-6-24-12-25)26-10-13-11-27-20-15(13)2-1-3-17(20)22;2-1(3)4/h1-8,11-12,19H,9-10H2;(H,2,3,4) |
| Løsningstype | Neat |
| SMIL | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3csc4c(Cl)cccc34)c(Cl)c1 |
| Vis mere |