Få mere at vide
Serricornin (Mixture of Isomers), TRC
Artikelnummer. 30133068
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133068 100 mg
30012177 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133068 Leverandør Toronto Research Chemicals Leverandørnr. TRCS278000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72522-40-8
Molekylær formel C11 H22 O2
Molekylvægt (g/mol) 186.29
Synonym Serricornin
IUPAC navn 7-hydroxy-4,6-dimethylnonan-3-one
SMIL CCC(O)C(C)CC(C)C(=O)CC
Tekniske data
Kemisk navn eller materiale Serricornin
Mængde 100 mg
InChI formel InChI=1S/C11H22O2/c1-5-10(12)8(3)7-9(4)11(13)6-2/h8-10,12H,5-7H2,1-4H3
Formel vægt 186.162