missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINB8 (Human) Recombinant Protein
Human SERPINB8 full-length ORF ( AAH34528, 1 a.a. - 242 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005271-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDDLCEANGTFAISLFKILGEEDNSRNVFFSPMSISSALAMVFMGAKGSTAAQMSQALCLYKDGDIHRGFQSLLSEVNRTGTQYLLRTANRLFGEKTCDFLPDFKEYCQKFYQAELEELSFAEDTEECRKHINDWVAEKTEGKISEVLDAGTVDPLTKLVLVNAIYFKGKWNEQFDRKYTRGMLFKTNEEKKTVQMMFKEAKFKMGYADEVHTQVLELPYVEEELSMVILLPDDNTDLAVKE
Specifications
AAH34528 | |
serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINB8 | |
GST |
5271 | |
Wheat germ expression system | |
10 μg | |
CAP2, PI8 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |