missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Serpin B8/Proteinase Inhibitor 8 Antibody (013), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90484IR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Mouse samples. It is validated for ELISA.
Tekniske data
| Serpin B8/Proteinase Inhibitor 8 | |
| Monoclonal | |
| DyLight 755 | |
| CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin B8/Proteinase Inhibitor 8 (NP_035589.1; Met 1-Pro 374). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 5271 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 13 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion