missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B5/Maspin Antibody (SERPINB5/4972), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14135R
Additional Details : Weight : 0.00970kg
Description
Serpin B5/Maspin Monoclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Serpin B5/Maspin | |
Monoclonal | |
DyLight 550 | |
maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
Recombinant fragment of human Serpin B5/Maspin protein (around aa1-200) (exact sequence is proprietary) (Uniprot: P36952) | |
0.1 mL | |
Biologically Active Proteins, Cancer, Tumor Suppressors | |
5268 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SERPINB5/4972 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |