missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Serine racemase Recombinant Rabbit Monoclonal Antibody (12B2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550071
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Serine racemase Recombinant Monoclonal Antibody for Western Blot, Flow, ELISA
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Tekniske data
| Serine racemase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9GZT4 | |
| Srr | |
| A synthesized peptide derived from human SRR. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Western Blot | |
| 12B2 | |
| Unconjugated | |
| Srr | |
| D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 63826 | |
| -20°C or -80°C if preferred | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion