Learn More
Hamilton™ Septa, High Temperature
Product Code. 10150601
Change view
Click to view available options
Quantity:
12/Pk.
Unit Size:
Pack of 12
Product Code. Quantity Unit Size
10150601 12/Pk. Pack of 12
1 options
Product Code. 10150601 Supplier Hamilton™ Supplier No. 75803
  • ONLINE EXCLUSIVE PRICE
    246.00 DKK
    221.55 DKK / Pakke med 12
    Spare 24.45 DKK
    9% Slukket
Forventet forsendelse: 09-12-2026
Specifications
Product Type Septum
Temperature (Metric) Max 212°C
Material PTFE-backed Silicone Rubber
Quantity 12/Pk.