Learn More
Thermo Scientific™ Septa for 8-425 Screw Thread Caps
Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
Brand: Thermo Scientific™ C4013-60TW
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
- PTFE Disk septa are chemically inert
- PFTE red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin gauge needles
Specifications
8 mm | |
PTFE, Silicone | |
8-425 |