Få mere at vide
SEPHS1 Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™
Artikelnummer. 30498880 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30498880 0.1 mL 0.1ml
1 options
Artikelnummer. 30498880 Leverandør Novus Biologicals Leverandørnr. NBP335339CL1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
SEPHS1 Polyclonal antibody specifically detects SEPHS1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen SEPHS1
Applikationer ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Klassifikation Polyclonal
Konjugeret CoraFluor 1
Formulering PBS
Gene Alias EC 2.7.9.3, MGC4980, SELD, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, selenophosphate synthetase 1 +E9a, selenophosphate synthetase 1 delta E2, selenophosphate synthetase 1 delta E8, selenophosphate synthetase 1 major, SPS1selenophosphate synthetase 1 +E9, water dikinase 1
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-392 of human SEPHS1 (NP_036379.2).,, Sequence:, MSTRESFNPESYELDKSFRLTRFTELKGTGCKVPQDVLQKLLESLQENHFQEDEQFLGAVMPRLGIGMDTCVIPLRHGGLSLVQTTDYIYPIVDDPYMMGRIACANVLSDLYAMGVTECDNMLMLLGVSNKMTDRERDKVMPLIIQGFKDAAEEAGTSVTGGQTVLNPWIVLGGVATTVCQPNEFIMPDNAVPGDVLVLTKPLGTQVAVAVHQWLDIPEKWNKIKLVVTQEDVELAYQEAMMNMARLNRTAAGLMHTFNAHAATDITGFGILGHAQNLAKQQRNEVSFVIHNLPVLAKMAAVSKACGNMFGLMHGTCPETSGGLLICLPREQAARFCAEIKSPKYGEGHQAWIIGIVEKGNRTARIIDKPRIIEVAPQVATQNVNPTPGATS
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Stem Cell Markers
Primær eller sekundær Primary
Gen-id (Entrez) 22929
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark. Do not freeze.
Produkttype Antibody
Form Purified
Isotype IgG