Learn More
Cytiva (formerly Pall Lab) Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filter Funnels are single-use funnels designed for use with the Sentino™ pump with a press-fit seal, membrane guides and forceps access point allowing for simple set up and membrane removal.
Brand: Cytiva (formerly Pall Lab) 4870
803.67 DKK valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 1.40000kg
Description
- For research use only
Specifications
100 mL | |
25/Sleeve, 100/Pk. | |
Filtering Funnel |
Polypropylene | |
Sterile |