Få mere at vide
Sennoside A, TRC
Artikelnummer. 30012730
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012730 250 mg
30009975 50 mg
30009652 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012730 Leverandør Toronto Research Chemicals Leverandørnr. TRCS258805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81-27-6
Molekylær formel C42 H38 O20
Molekylvægt (g/mol) 862.74
Synonym [9,9'-Bianthracene]-2,2'-dicarboxylic acid, 5,5'-bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro-4,4'-dihydroxy-10,10'-dioxo-, (9R,9'R)-, Sennoside A (6CI,7CI,8CI), [9,9'-Bianthracene]-2,2'-dicarboxylic acid, 5,5'-bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro-4,4'-dihydroxy-10,10'-dioxo-, (R*,R*)-, rel-(9R,9'R)-5,5'-Bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro-4,4'-dihydroxy-10,10'-dioxo[9,9'-bianthracene]-2,2'-dicarboxylic acid, Gentle Nature, Glysennid, NSC 112929, (9R,9'R)-5,5'-Bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro-4,4'-dihydroxy-10,10'-dioxo[9,9'-bianthracene]-2,2'-dicarboxylic acid
IUPAC navn (9R)-9-[(9R)-2-carboxy-4-hydroxy-10-oxo-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-9H-anthracen-9-yl]-4-hydroxy-10-oxo-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-9H-anthracene-2-carboxylic acid
SMIL OC[C@H]1O[C@@H](Oc2cccc3[C@@H]([C@@H]4c5cccc(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(=O)O)c8cc(cc(O)c8C(=O)c23)C(=O)O)[C@H](O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Sennoside A
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C42H38O20/c43-11-23-31(47)35(51)37(53)41(61-23)59-21-5-1-3-15-25(17-7-13(39(55)56)9-19(45)27(17)33(49)29(15)21)26-16-4-2-6-22(60-42-38(54)36(52)32(48)24(12-44)62-42)30(16)34(50)28-18(26)8-14(40(57)58)10-20(28)46/h1-10,23-26,31-32,35-38,41-48,51-54H,11-12H2,(H,55,56)(H,57,58)/t23-,24-,25-,26-,31-,32-,35+,36+,37-,38-,41-,42-/m1/s1
Formel vægt 862.1956
Procent renhed >95%