Learn More
Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves
Disposable protective glove made of Nitrile. Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves are latex free with high wearing comfort.
Brand:  Semperit™ 3000004106
Additional Details : Weight : 0.47250kg
Product Code. 15553546
  • 24.24 DKK / Pack of 100
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Description
Description
  • CE Category III protective glove for complex risks. CE Class I examination glove.
  • Special nitrile formulation with optimized resistance to oils and fats, good tear-resistance
  • Excellent wearing comfort
  • Tested and certified according to 1935/2004 - suitable for food contact
  • The problem-solver for single use to protect hands against chemical splashing and in the processing of fatty foods.
  • Free from phthalates / softeners and allergenic latex proteins
  • Signal color blue – suitable for areas subject to HACCP standards