missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ SEMA4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101258
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total SEMA4A.
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.
Tekniske data
| SEMA4A | |
| Polyclonal | |
| Unconjugated | |
| SEMA4A | |
| AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20351, 310630, 64218 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q62178, Q9H3S1 | |
| SEMA4A | |
| A synthesized peptide derived from human SEMA4A(Accession Q9H3S1), corresponding to amino acid residues P512-P562. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion