missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELM Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00138B
Additional Details : Weight : 0.00970kg
Description
SELM Polyclonal specifically detects SELM in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SELM | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC40146, selenoprotein M, selenoprotein SelM, SEPM | |
Produced in rabbits immunized with purified, recombinant SELM (Q8WWX9; Ala24-Leu145). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
140606 | |
Store at 4°C in the dark. | |
IgG |