Få mere at vide
Globe Scientific Self-Standing Transport Tubes with Attached Screw Cap
Shop alle Globe Scientific produkterBeskrivelse
- Tubes are made of polypropylene
- With molded graduations
- Self-standing with conical bottom
- Transparent tubes allows for unobstructed viewing of samples
- Shatter-proof and may be exposed to freezing temperatures
- Leak-resistant polyethylene screw caps can be removed or sealed in single turn
Tekniske data
Tekniske data
| Lukningsfarve | Hvid |
| Autoklaverbar | Autoklaverbar (kun flaske) |
| Kapacitet (metrisk) | 5 mL |
| Materiale | Polypropylen |
| Gradueringer | 1 til 5 ml |
| Inkluderer | Hvid skruedæksel |
| Stil | Selvstående transportrør |
| Sterilitet | Ikke steril |
| Engangs | Engangsbrug |
| Uddannet | Ja |
| Vis mere |