Få mere at vide
Globe Scientific Selvstående transportrør
Artikelnummer. 31923634 Shop alle Globe Scientific produkter
Skift visning
Kapacitet (metrisk):
5 mL
10 mL
Farve:
Amber
Clear
Lukning inkluderet:
No
Yes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapacitet (metrisk) Farve Lukning inkluderet
31923634 5 mL Clear No
31923153 5 mL Amber Yes
31921775 10 mL Clear Yes
31922351 10 mL Clear No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 31923634 Leverandør Globe Scientific Leverandørnr. 6101TUBE
Forventet forsendelse: 11-11-2026
Designet til sikker opbevaring og transport af biologiske materialer til 6101- eller 6102-seriens transportrør
  • The transparent polypropylene (PP) vials allow unobstructed viewing of the sample
  • The vials are shatter-proof and may be exposed to freezing temperatures
  • They feature leak-resistant screw caps that can be removed or sealed in a single turn
Tekniske data
Farve Clear
Autoklaverbar Autoclavable
Kapacitet (metrisk) 5 mL
Bund form Conical (Self-standing)
Materiale Polypropylene
Trådstil External
Sterilitet Non-sterile
Uddannet Yes
Lukning inkluderet No
Produkttype Transport Tube (Self-Standing)