Få mere at vide
Globe Scientific Self-Standing Transport Tubes
Artikelnummer. 31922655 Shop alle Globe Scientific produkter
Skift visning
Kapacitet (metrisk):
10 mL
Artikelnummer. Kapacitet (metrisk)
31922655 10 mL
1 options
Artikelnummer. 31922655 Leverandør Globe Scientific Leverandørnr. 6102AM
Forventet forsendelse: 11-11-2026
Globe Scientific Inc. Self-Standing Transport Tubes with leak-resistant screw caps
  • Designed for the safe storage and transport of biological materials
  • Transparent polypropylene (PP) vials allow unobstructed viewing of the sample
  • Vials are shatter-proof and may be exposed to freezing temperatures
  • Leak-resistant screw caps can be removed or sealed with a single turn
Tekniske data
Lukningsfarve White
Lukningsmateriale Polyethylene
Farve Amber
Kapacitet (metrisk) 10 mL
Længde (metrisk) 92 mm
Materiale Polypropylene
Engangs Single-use
Sterilitet Non-sterile
Autoklaverbar Autoclavable (Without Cap)
Uddannet Yes
Diameter (metrisk) 16 mm
Lukketype Screw Cap
Produkttype Transport Tube