missing translation for 'onlineSavingsMsg'
Learn More

Selenourea, 99%, Thermo Scientific Chemicals

Catalog Number p-7050405
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
630-10-4
CH3N2Se
122.02
MFCD00008065
ZUCFIOXGLBMWHV-UHFFFAOYSA-N
selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
6327594
$l^{1}-selanylmethanimidamide
C(=N)(N)[Se]
This item is not returnable. View return policy
630-10-4
CH3N2Se
122.02
MFCD00008065
ZUCFIOXGLBMWHV-UHFFFAOYSA-N
selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
6327594
$l^{1}-selanylmethanimidamide
C(=N)(N)[Se]

Selenourea is used in the synthesis of selenium heterocycles, which exhibit anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point ∼210°C (decomposition)
Color White to Gray
Packaging Ampule
Quantity 1 g
UN Number UN3283
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 123.02
Percent Purity 99.97%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Selenourea
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'